2,4-Dimethoxy-5-nitropyrimidine 97%
Supplier: Ambeed
|
Warning
|
A103264-100G
A103264-10G
A103264-1G
A103264-250MG
A103264-25G
A103264-5G
77188-640EA
814.73
USD
77188-640
77188-642
77188-644
77188-646
77188-648
77188-650
2,4-Dimethoxy-5-nitropyrimidine 97%
2,4-Dimethoxy-5-nitropyrimidine
Precaución: for Research Use Only
Formula:
C₆H₇N₃O₄ MW: 185.14 g/mol |
MDL Number:
MFCD00726922 CAS Number: 30561-07-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...