Benzofuro[3,2-d]pyrimidine-2,4(1H,3H)-dione 95%

Supplier: Ambeed
Warning

A103238-1G A103238-250MG A103238-5G
77160-322EA 34 USD
77160-322 77160-324 77160-326
Benzofuro[3,2-d]pyrimidine-2,4(1H,3H)-dione 95%
Benzofuro[3,2-d]pyrimidine-2,4(1H,3H)-dione

Precaución: for Research Use Only

Formula: C₁₀H₆N₂O₃
MDL Number: MFCD00788168
CAS Number: 62208-68-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR