(2S,3S)-2,3-Bis((2-methylbenzoyl)oxy)succinic acid 95%

Supplier: Ambeed
Warning

A103002-100MG A103002-1G A103002-250MG A103002-5G
77160-310EA 27.81 USD
77160-310 77160-312 77160-314 77565-780
(2S,3S)-2,3-Bis((2-methylbenzoyl)oxy)succinic acid 95%
(2S,3S)-2,3-Bis((2-methylbenzoyl)oxy)succinic acid

Precaución: For research use only.

Formula: C₂₀H₁₈O₈
MW: 386.35 g/mol
MDL Number: MFCD11114441
CAS Number: 391624-66-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR