1-Bromo-4-chloro-2,5-difluorobenzene 97%

Supplier: Ambeed
Warning

A102954-10G A102954-250MG A102954-1G A102954-5G
77160-306EA 55 USD
77160-306 77160-308 77891-759 77891-760
1-Bromo-4-chloro-2,5-difluorobenzene 97%
1-Bromo-4-chloro-2,5-difluorobenzene

Precaución: for Research Use Only

Formula: C₆H₂BrClF₂
MDL Number: MFCD09878116
CAS Number: 172921-33-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR