(S)-1-tert-Butyl-2-methyl-4-oxopiperidine-1,2-dicarboxylate 97%

Supplier: Ambeed
Warning

Synonyms: (S)-1-tert-Butyl 2-methyl 4-oxopiperidine-1,2-dicarboxylate

A102660-100MG A102660-1G A102660-250MG A102660-25G A102660-5G
77160-288EA 40.17 USD
77160-288 77160-290 77160-292 77160-294 77160-296
(S)-1-tert-Butyl-2-methyl-4-oxopiperidine-1,2-dicarboxylate 97%
(S)-1-tert-Butyl-2-methyl-4-oxopiperidine-1,2-dicarboxylate

Precaución: for Research Use Only

Formula: C₁₂H₁₉NO₅
MW: 257.29 g/mol
MDL Number: MFCD12547027
CAS Number: 756486-14-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR