3-Amino-1-methylpyridin-2(1H)-one hydrochloride 98%
Supplier: Ambeed
|
Warning
|
A102611-1G
A102611-250MG
A102611-5G
77160-282EA
57
USD
77160-282
77160-284
77160-286
3-Amino-1-methylpyridin-2(1H)-one hydrochloride 98%
3-Amino-1-methylpyridin-2(1H)-one hydrochloride
Precaución: for Research Use Only
Formula:
C₆H₉ClN₂O |
MDL Number:
MFCD27986870 CAS Number: 1523570-95-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...