1-Boc-5-cyanoindole 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
tert-Butyl 5-cyano-1H-indole-1-carboxylate
A102574-1G
A102574-25G
A102574-5G
77188-620EA
11.33
USD
77188-620
77188-622
77188-624
1-Boc-5-cyanoindole 98%
1-Boc-5-cyanoindole
Precaución: for Research Use Only
Formula:
C₁₄H₁₄N₂O₂ MW: 242.28 g/mol |
MDL Number:
MFCD08056374 CAS Number: 475102-10-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...