4-Chloro-6-methoxyquinazoline 95%

Supplier: Ambeed
Warning

A102404-100MG A102404-10G A102404-250MG A102404-1G
77160-272EA 14 USD
77160-272 77160-274 77160-276 77610-766
4-Chloro-6-methoxyquinazoline 95%
4-Chloro-6-methoxyquinazoline

Precaución: for Research Use Only

Formula: C₉H₇ClN₂O
MDL Number: MFCD06657615
CAS Number: 50424-28-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR