1,5-Diiodopentane 97% stabilized
Supplier: Ambeed
|
Warning
|
Synonyms:
Pentamethylene diiodide
Stabilization:
Stabilized with Copper chip
A102343-100G
A102343-10G
A102343-1G
A102343-25G
A102343-5G
A102343-500G
77188-608EA
125.66
USD
77188-608
77188-610
77188-612
77188-614
77188-616
77745-780
1,5-Diiodopentane 97% stabilized
1,5-Diiodopentane
Precaución: for Research Use Only
Formula:
C₅H₁₀I₂ MW: 323.94 g/mol |
MDL Number:
MFCD00001101 CAS Number: 628-77-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...