(2-(tert-Butoxycarbonyl)-1,2,3,4-tetrahydroisoquinolin-6-yl)boronic acid 98%
Supplier: Ambeed
|
Warning
|
A102332-100MG
A102332-1G
A102332-250MG
A102332-5G
77160-262EA
88
USD
77160-262
77160-264
77160-266
77160-268
(2-(tert-Butoxycarbonyl)-1,2,3,4-tetrahydroisoquinolin-6-yl)boronic acid 98%
(2-(tert-Butoxycarbonyl)-1,2,3,4-tetrahydroisoquinolin-6-yl)boronic acid
Precaución: for Research Use Only
Formula:
C₁₄H₂₀BNO₄ |
MDL Number:
MFCD08752239 CAS Number: 1312765-94-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...