N-(Pyrazinylcarbonyl)-L-phenylalanine 98%

Supplier: Ambeed
Warning

A102305-100G A102305-1G A102305-250MG A102305-5G A102305-25G
77188-600EA 227 USD
77188-600 77188-602 77188-604 77188-606 77745-779
N-(Pyrazinylcarbonyl)-L-phenylalanine 98%
N-(Pyrazinylcarbonyl)-L-phenylalanine

Precaución: for Research Use Only

Formula: C₁₄H₁₃N₃O₃
MDL Number: MFCD09800536
CAS Number: 114457-94-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR