Styrene-4-sulfonic acid sodium salt 90%, Ambeed.Inc

Supplier: Ambeed
Warning

Synonyms: Sodium 4-vinylbenzenesulphonate, 4-Vinylbenzenesulphonic acid sodium salt, Sodium-p-styrenesulphonate, p-Styrenesulphonic acid sodium salt

A102096-100G A102096-10G A102096-25G A102096-500G A102096-1000G
77160-242EA 9.27 USD
77160-242 77160-244 77160-246 77160-248 77682-939
Styrene-4-sulfonic acid sodium salt 90%, Ambeed.Inc
Styrene-4-sulfonic acid sodium salt

Precaución: For research use only.

Formula: C₈H₇NaO₃S
MW: 206.19 g/mol
MDL Number: MFCD00013379
CAS Number: 2695-37-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR