5-Amino-2,3-dihydro-2-methyl-1H-isoindol-1-one 98%, Ambeed.Inc
Supplier: Ambeed
|
Warning
|
Synonyms:
5-Amino-2-methylisoindolin-1-one
A101814-1G
A101814-5G
A101814-250MG
77160-216EA
112.27
USD
77160-216
77160-218
77682-938
5-Amino-2,3-dihydro-2-methyl-1H-isoindol-1-one 98%, Ambeed.Inc
5-Amino-2,3-dihydro-2-methyl-1H-isoindol-1-one
Precaución: For research use only.
Formula:
C₉H₁₀N₂O MW: 162.19 g/mol |
MDL Number:
MFCD17214360 CAS Number: 1190380-38-1 |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...