(5-Methylbenzofuran-2-yl)boronic acid 98%

Supplier: Ambeed
Warning

A101670-100MG A101670-250MG A101670-1G
77160-208EA 60 USD
77160-208 77548-916 77891-752
(5-Methylbenzofuran-2-yl)boronic acid 98%
(5-Methylbenzofuran-2-yl)boronic acid

Precaución: for Research Use Only

Formula: C₉H₉BO₃
MDL Number: MFCD28101768
CAS Number: 143381-72-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR