2-(4-Bromo-3-methoxyphenyl)acetonitrile 98%

Supplier: Ambeed
Warning

A101535-1G A101535-250MG
77160-204EA 60.77 USD
77160-204 77160-206
2-(4-Bromo-3-methoxyphenyl)acetonitrile 98%
2-(4-Bromo-3-methoxyphenyl)acetonitrile

Precaución: for Research Use Only

Formula: C₉H₈BrNO
MW: 226.06 g/mol
MDL Number: MFCD06797367
CAS Number: 113081-50-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR