1,2-Dimyristoyl-sn-glycero-3-phosphocholine 98%

Supplier: Ambeed
Warning

Synonyms: 14:0 PC, 1,2-Ditetradecanoyl-sn-glycero-3-phosphocholine, (R)-2,3-Bis(tetradecanoyloxy)propyl (2-(trimethylammonio)ethyl) phosphate, DMPC, β-γ-Dimyristoyl-L-α-Lecithin, (R)-2,3-Bis(tetradecanoyloxy)propyl [2-(Trimethylammonio)ethyl] Phosphate

A101534-5G A101534-100MG A101534-1G A101534-250MG A101534-100G A101534-25G
77188-564EA 561.35 USD
77188-564 77614-746 77614-748 77614-750 77745-774 77745-775
1,2-Dimyristoyl-sn-glycero-3-phosphocholine 98%
1,2-Dimyristoyl-sn-glycero-3-phosphocholine

Precaución: for Research Use Only

Formula: C₃₆H₇₂NO₈P
MW: 677.94 g/mol
MDL Number: MFCD00036901
CAS Number: 18194-24-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR