2-Amino-3,5-difluorobenzonitrile 98%

Supplier: Ambeed
Warning

A100317-1G A100317-250MG A100317-5G
77160-150EA 20 USD
77160-150 77160-152 77160-154
2-Amino-3,5-difluorobenzonitrile 98%
2-Amino-3,5-difluorobenzonitrile

Precaución: for Research Use Only

Formula: C₇H₄F₂N₂
MDL Number: MFCD18826239
CAS Number: 126674-94-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR