(R)-Piperidine-3-carboxylic acid amide 97%

Supplier: Ambeed
Warning

A999798-1G A999798-250MG A999798-5G
77160-136EA 54.59 USD
77160-136 77160-138 77160-140
(R)-Piperidine-3-carboxylic acid amide 97%
(R)-Piperidine-3-carboxylic acid amide

Precaución: for Research Use Only

Formula: C₆H₁₂N₂O
MW: 128.17 g/mol
MDL Number: MFCD17015910
CAS Number: 168749-30-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR