3-Chloro-4-fluorophenylacetonitrile 98%

Supplier: Ambeed
Warning

A998580-10G A998580-1G
77160-132EA 41.2 USD
77160-132 77160-134
3-Chloro-4-fluorophenylacetonitrile 98%
3-Chloro-4-fluorophenylacetonitrile

Precaución: for Research Use Only

Formula: C₈H₅ClFN
MW: 169.5834 g/mol
MDL Number: MFCD01631553
CAS Number: 658-98-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR