2-Methyl-2H-chromene-3-carbaldehyde 95%

Supplier: Ambeed
Warning

A997001-1G A997001-100MG A997001-250MG
77160-130EA 290.01 USD
77160-130 77900-929 77901-894
2-Methyl-2H-chromene-3-carbaldehyde 95%
2-Methyl-2H-chromene-3-carbaldehyde

Precaución: for Research Use Only

Formula: C₁₁H₁₀O₂
MDL Number: MFCD01662425
CAS Number: 57543-42-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR