2-Chloro-6,7-dihydro-5H-pyrrolo[3,4-b]pyridin-5-one 95%

Supplier: Ambeed
Warning

A995706-1G A995706-5G
77160-128EA 346.08 USD
77160-128 77687-013
2-Chloro-6,7-dihydro-5H-pyrrolo[3,4-b]pyridin-5-one 95%
2-Chloro-6,7-dihydro-5H-pyrrolo[3,4-b]pyridin-5-one

Precaución: for Research Use Only

Formula: C₇H₅ClN₂O
MW: 168.58 g/mol
MDL Number: MFCD18256428
CAS Number: 1256811-82-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR