Cyclopropyl-3-methylphenyl ketone 95%

Supplier: Ambeed
Warning

A995654-100MG A995654-1G A995654-250MG
77160-122EA 19 USD
77160-122 77160-124 77160-126
Cyclopropyl-3-methylphenyl ketone 95%
Cyclopropyl-3-methylphenyl ketone

Precaución: for Research Use Only

Formula: C₁₁H₁₂O
MDL Number: MFCD03841207
CAS Number: 150668-37-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR