(2R,4R)-tert-Butyl 4-hydroxy-2-methylpiperidine-1-carboxylate 98%

Supplier: Ambeed
Warning

A990253-100MG A990253-1G A990253-250MG A990253-5G
77160-116EA 47.38 USD
77160-116 77160-118 77160-120 77614-712
(2R,4R)-tert-Butyl 4-hydroxy-2-methylpiperidine-1-carboxylate 98%
(2R,4R)-tert-Butyl 4-hydroxy-2-methylpiperidine-1-carboxylate

Precaución: for Research Use Only

Formula: C₁₁H₂₁NO₃
MW: 215.29 g/mol
MDL Number: MFCD23105950
CAS Number: 790667-44-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR