1-Butyl-4-methylpyridiniumhexafluorophosphate 97%

Supplier: Ambeed
Warning

A988776-100G A988776-25G A988776-500G A988776-5G
77160-104EA 79 USD
77160-104 77160-106 77160-108 77160-110
1-Butyl-4-methylpyridiniumhexafluorophosphate 97%
1-Butyl-4-methylpyridiniumhexafluorophosphate

Precaución: for Research Use Only

Formula: C₁₀H₁₆F₆NP
MDL Number: MFCD03095438
CAS Number: 401788-99-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR