2,3,6-Trifluorophenol 95%

Supplier: Ambeed
Warning

A984572-10G A984572-25G A984572-5G
77160-096EA 49 USD
77160-096 77160-098 77160-100
2,3,6-Trifluorophenol 95%
2,3,6-Trifluorophenol

Precaución: for Research Use Only

Formula: C₆H₃F₃O
MDL Number: MFCD00061214
CAS Number: 113798-74-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR