2,3,6-Trifluorobenzonitrile 97%

Supplier: Ambeed
Danger

A981418-1G A981418-250MG A981418-25G A981418-5G
77160-084EA 31.93 USD
77160-084 77160-086 77160-088 77160-090
2,3,6-Trifluorobenzonitrile 97%
2,3,6-Trifluorobenzonitrile

Precaución: for Research Use Only

Formula: C₇H₂F₃N
MW: 157.09 g/mol
MDL Number: MFCD00012287
CAS Number: 136514-17-5
UN: 3276
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR