(6-(Difluoromethoxy)pyridin-3-yl)boronic acid 95%

Supplier: Ambeed
Warning

A975440-1G A975440-250MG A975440-100MG
77160-080EA 491 USD
77160-080 77160-082 77901-690
(6-(Difluoromethoxy)pyridin-3-yl)boronic acid 95%
(6-(Difluoromethoxy)pyridin-3-yl)boronic acid

Precaución: for Research Use Only

Formula: C₆H₆BF₂NO₃
MDL Number: MFCD18261896
CAS Number: 1354290-88-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR