Potassium trifluoro(5-methylfuran-2-yl)borate 98%

Supplier: Ambeed
Warning

A969335-10G A969335-25G A969335-5G
77160-052EA 47.38 USD
77160-052 77160-054 77160-056
Potassium trifluoro(5-methylfuran-2-yl)borate 98%
Potassium trifluoro(5-methylfuran-2-yl)borate

Precaución: for Research Use Only

Formula: C₅H₅BF₃KO
MW: 187.99 g/mol
MDL Number: MFCD09992923
CAS Number: 1111213-54-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR