1-[2-Oxo-2-(2-pyridyl)ethyl]pyridinium iodide 98%

Supplier: Ambeed
Warning

Synonyms: 1-(2-Oxo-2-(pyridin-2-yl)ethyl)pyridin-1-ium iodide

A968884-1G A968884-25G A968884-5G
77160-046EA 24.72 USD
77160-046 77160-048 77160-050
1-[2-Oxo-2-(2-pyridyl)ethyl]pyridinium iodide 98%
1-[2-Oxo-2-(2-pyridyl)ethyl]pyridinium iodide

Precaución: for Research Use Only

Formula: C₁₂H₁₁IN₂O
MW: 326.14 g/mol
MDL Number: MFCD01690884
CAS Number: 26482-00-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR