3-Bromo-5-isopropylbenzoic acid 96%

Supplier: Ambeed
Warning

A968637-1G A968637-100MG A968637-250MG
77160-044EA 51 USD
77160-044 77901-807 77901-397
3-Bromo-5-isopropylbenzoic acid 96%
3-Bromo-5-isopropylbenzoic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₁BrO₂
MDL Number: MFCD20040502
CAS Number: 112930-39-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR