1,2,3,4,6-Penta-O-acetyl-D-mannopyranose 97%

Supplier: Ambeed
Warning

Synonyms: alpha,beta-D-Mannose pentaacetate, 1,2,3,4,6-Penta-O-acetyl-D-mannopyranose, (3S,4S,5R,6R)-6-(Acetoxymethyl)tetrahydro-2H-pyran-2,3,4,5-tetrayl tetraacetate

A967369-100G A967369-1G A967369-5G A967369-25G A967369-500G
77160-038EA 147 USD
77160-038 77160-040 77160-042 77614-688 77614-690
1,2,3,4,6-Penta-O-acetyl-D-mannopyranose 97%
1,2,3,4,6-Penta-O-acetyl-D-mannopyranose

Precaución: for Research Use Only

Formula: C₁₆H₂₂O₁₁
MDL Number: MFCD05864874
CAS Number: 25941-03-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR