2-(3-Bromophenyl)triphenylene 98%

Supplier: Ambeed
Warning

A964564-250MG A964564-25G A964564-5G A964564-1G
77160-032EA 9.27 USD
77160-032 77160-034 77160-036 77686-991
2-(3-Bromophenyl)triphenylene 98%
2-(3-Bromophenyl)triphenylene

Precaución: for Research Use Only

Formula: C₂₄H₁₅Br
MW: 383.29 g/mol
MDL Number: MFCD28384139
CAS Number: 1313514-53-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR