TD-139 95+%

Supplier: Ambeed
Warning

Synonyms: TD139, (2R,2'R,3R,3'R,4S,4'S,5R,5'R,6S,6'S)-6,6'-Thiobis(4-(4-(3-fluorophenyl)-1H-1,2,3-triazol-1-yl)-2-(hydroxymethyl)tetrahydro-2H-pyran-3,5-diol), (2S,3R,4S,5R,6R)-4-[4-(3-fluorophenyl)triazol-1-yl]-2-[(2S,3R,4S,5R,6R)-4-[4-(3-fluorophenyl)triazol-1-yl]-3,5-dihydroxy-6-(hydroxymethyl)oxan-2-yl]sulphanyl-6-(hydroxymethyl)oxane-3,5-diol

A955559-1MG A955559-5MG
77160-016EA 33.99 USD
77160-016 77160-018
TD-139 95+%
TD-139

Precaución: for Research Use Only

Formula: C₂₈H₃₀F₂N₆O₈S
MW: 648.64 g/mol
MDL Number: MFCD29919388
CAS Number: 1450824-22-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR