Tetrabutylammonium tetraphenylborate 98%

Supplier: Ambeed
Warning

Synonyms: Tetrabutylammonium Tetraphenylborate

A954617-1G A954617-5G A954617-25G
77160-012EA 19.57 USD
77160-012 77160-014 77552-772
Tetrabutylammonium tetraphenylborate 98%
Tetrabutylammonium tetraphenylborate

Precaución: for Research Use Only

Formula: C₄₀H₅₆BN
MW: 561.6904 g/mol
MDL Number: MFCD00011749
CAS Number: 15522-59-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR