1-(2-Methoxyethyl)piperidin-4-ol 98+%
Supplier: Ambeed
|
Warning
|
A951835-1G
A951835-250MG
A951835-5G
77159-994EA
41
USD
77159-994
77159-996
77900-930
1-(2-Methoxyethyl)piperidin-4-ol 98+%
1-(2-Methoxyethyl)piperidin-4-ol
Precaución: for Research Use Only
Formula:
C₈H₁₇NO₂ |
MDL Number:
MFCD13178921 CAS Number: 512778-95-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...