Bis(2,4-pentanedionato)tin(IV) dichloride 98+%

Supplier: Ambeed
Danger

Synonyms: Tin(IV) Acetylacetonate Dichloride, Tin(IV) Chloride Bis(2,4-pentanedionate), Acetylacetone Tin(IV) Dichloride Salt

A951477-1G A951477-25G A951477-5G
77159-988EA 14.42 USD
77159-988 77159-990 77159-992
Bis(2,4-pentanedionato)tin(IV) dichloride 98+%
Bis(2,4-pentanedionato)tin(IV) dichloride

Precaución: for Research Use Only

Formula: C₁₀H₁₄Cl₂O₄Sn
MW: 387.83 g/mol
MDL Number: MFCD00015316
CAS Number: 16919-46-3
UN: 3261
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR