(E)-2-(3-(3,4-Dimethoxyphenyl)acrylamido)benzoic acid 98%
Supplier: Ambeed
|
Warning
|
A941193-100MG
A941193-10MG
A941193-1G
A941193-1MG
A941193-250MG
A941193-25MG
A941193-50MG
A941193-5G
77159-948EA
36
USD
77159-948
77159-950
77159-952
77159-954
77159-956
77159-958
77159-960
77159-962
(E)-2-(3-(3,4-Dimethoxyphenyl)acrylamido)benzoic acid 98%
(E)-2-(3-(3,4-Dimethoxyphenyl)acrylamido)benzoic acid
Precaución: for Research Use Only
Formula:
C₁₈H₁₇NO₅ |
MDL Number:
MFCD00864787 CAS Number: 70806-55-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...