5,6-Dichlorobenzimidazole-1-β-D-Ribofuranoside 98%

Supplier: Ambeed
Warning

Synonyms: 5,6-Dichloro-1-(β-D-ribofuranosyl)benzimidazole, 5,6-Dichlorobenzimidazole Riboside, (2R,3R,4S,5R)-2-(5,6-Dichloro-1H-benzo[d]imidazol-1-yl)-5-(hydroxymethyl)tetrahydrofuran-3,4-diol, DRB

A941159-100MG A941159-250MG
77159-946EA 108.15 USD
77159-946 77686-968
5,6-Dichlorobenzimidazole-1-β-D-Ribofuranoside 98%
5,6-Dichlorobenzimidazole-1-beta-D-Ribofuranoside

Precaución: for Research Use Only

Formula: C₁₂H₁₂Cl₂N₂O₄
MW: 319.14 g/mol
MDL Number: MFCD00036785
CAS Number: 53-85-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR