Methyl cis-4-hydroxycyclohexanecarboxylate 98%

Supplier: Ambeed
Warning

Synonyms: cis-Methyl 4-hydroxycyclohexanecarboxylate

A938533-1G A938533-5G
77159-942EA 32.96 USD
77159-942 77159-944
Methyl cis-4-hydroxycyclohexanecarboxylate 98%
Methyl cis-4-hydroxycyclohexanecarboxylate

Precaución: for Research Use Only

Formula: C₈H₁₄O₃
MW: 158.197 g/mol
MDL Number: MFCD20441962
CAS Number: 3618-03-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR