2-Chloro-4-ethoxy-5-methoxypyrimidine 95%

Supplier: Ambeed
Warning

A938428-100MG A938428-250MG
77159-938EA 82.4 USD
77159-938 77159-940
2-Chloro-4-ethoxy-5-methoxypyrimidine 95%
2-Chloro-4-ethoxy-5-methoxypyrimidine

Precaución: for Research Use Only

Formula: C₇H₉ClN₂O₂
MW: 188.61 g/mol
MDL Number: MFCD21098378
CAS Number: 1353976-54-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR