5-Chloro-1-(4-methoxyphenyl)-1-oxopentane 95%

Supplier: Ambeed
Warning

A932109-5G A932109-1G
77159-934EA 605.64 USD
77159-934 77686-966
5-Chloro-1-(4-methoxyphenyl)-1-oxopentane 95%
5-Chloro-1-(4-methoxyphenyl)-1-oxopentane

Precaución: for Research Use Only

Formula: C₁₂H₁₅ClO₂
MDL Number: MFCD00517502
CAS Number: 949-06-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR