Levocetirizine dihydrochloride 98%

Supplier: Ambeed
Danger

A930064-25G A930064-25MG A930064-5G A930064-1MG A930064-5MG
77159-920EA 55 USD
77159-920 77159-922 77159-924 77686-964 77686-965
Levocetirizine dihydrochloride 98%
Levocetirizine dihydrochloride

Precaución: for Research Use Only

Formula: C₂₁H₂₅ClN₂O₃
MDL Number: MFCD07366507
CAS Number: 130018-87-0
UN: 3077
ADR: 9,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR