3,5-Difluorobenzyl bromide 98%

Supplier: Ambeed
Danger

Synonyms: α-Bromo-3,5-difluorotoluene

A929733-100G A929733-10G A929733-1G A929733-25G A929733-5G
77159-910EA 163 USD
77159-910 77159-912 77159-914 77159-916 77159-918
3,5-Difluorobenzyl bromide 98%
3,5-Difluorobenzyl bromide

Precaución: for Research Use Only

Formula: C₇H₅BrF₂
MDL Number: MFCD00010304
CAS Number: 141776-91-2
UN: 3265
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR