tert-Butyl 4-((4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)methylene)piperidine-1-carboxylate 98%

Supplier: Ambeed
Warning

A925071-1G A925071-250MG A925071-25G A925071-5G A925071-100MG
77159-902EA 104.03 USD
77159-902 77159-904 77159-906 77159-908 77614-644
tert-Butyl 4-((4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)methylene)piperidine-1-carboxylate 98%
tert-Butyl 4-((4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)methylene)piperidine-1-carboxylate

Precaución: for Research Use Only

Formula: C₁₇H₃₀BNO₄
MW: 323.23 g/mol
MDL Number: MFCD14706665
CAS Number: 1425970-61-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR