3-Bromo-5-chlorobenzyl alcohol 98%

Supplier: Ambeed
Warning

Synonyms: (3-Bromo-5-chlorophenyl)methanol

A922768-250MG A922768-1G A922768-5G
77159-898EA 32.96 USD
77159-898 77686-956 77686-957
3-Bromo-5-chlorobenzyl alcohol 98%
3-Bromo-5-chlorobenzyl alcohol

Precaución: for Research Use Only

Formula: C₇H₆BrClO
MW: 221.479 g/mol
MDL Number: MFCD08061830
CAS Number: 917562-09-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR