Methyl 2-(3-bromophenyl)propanoate 95%

Supplier: Ambeed
Warning

A919296-100MG A919296-250MG A919296-5G
77159-888EA 54.59 USD
77159-888 77159-890 77159-892
Methyl 2-(3-bromophenyl)propanoate 95%
Methyl 2-(3-bromophenyl)propanoate

Precaución: for Research Use Only

Formula: C₁₀H₁₁BrO₂
MW: 243.10 g/mol
MDL Number: MFCD18908974
CAS Number: 80622-53-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR