7-Bromo-2,4-dichloropyrido[3,2-d]pyrimidine 95%

Supplier: Ambeed
Warning

A914003-100MG A914003-1G A914003-250MG
77159-866EA 78.28 USD
77159-866 77159-868 77159-870
7-Bromo-2,4-dichloropyrido[3,2-d]pyrimidine 95%
7-Bromo-2,4-dichloropyrido[3,2-d]pyrimidine

Precaución: for Research Use Only

Formula: C₇H₂BrCl₂N₃
MW: 278.92 g/mol
MDL Number: MFCD13250074
CAS Number: 1215074-41-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR