2-(3,5-Bis(trifluoromethyl)phenyl)-N,2-dimethyl-N-(6-(4-methylpiperazin-1-yl)-4-(o-tolyl)pyridin-3-yl)propanamide 98%

Supplier: Ambeed
Warning

Synonyms: Netupitant

A913871-10MG A913871-25MG A913871-5MG A913871-250MG A913871-5G
77159-862EA 5.15 USD
77159-862 77159-864 77595-474 77686-949 77686-950
2-(3,5-Bis(trifluoromethyl)phenyl)-N,2-dimethyl-N-(6-(4-methylpiperazin-1-yl)-4-(o-tolyl)pyridin-3-yl)propanamide 98%
2-(3,5-Bis(trifluoromethyl)phenyl)-N,2-dimethyl-N-(6-(4-methylpiperazin-1-yl)-4-(o-tolyl)pyridin-3-yl)propanamide

Precaución: for Research Use Only

Formula: C₃₀H₃₂F₆N₄O
MW: 578.59 g/mol
MDL Number: MFCD25976831
CAS Number: 290297-26-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR