2-Hydroxypyridine 98%
Supplier: Ambeed
|
Danger
|
Synonyms:
Pyridin-2-one, Pyridin-2(1H)-one, α-Hydroxypyridine, 2-Pyridone, α-Pyridone
A913349-10G
A913349-25G
A913349-5G
A913349-100G
77159-856EA
6.18
USD
77159-856
77159-858
77159-860
77614-634
2-Hydroxypyridine 98%
2-Hydroxypyridine
Precaución: for Research Use Only
Formula:
C₅H₅NO MW: 95.10 g/mol |
MDL Number:
MFCD00006268 CAS Number: 142-08-5 UN: 2811 ADR: 6.1,Ⅲ |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...