(4-Bromobenzyl)hydrazine hydrochloride 95%

Supplier: Ambeed
Warning

A913345-1G A913345-250MG A913345-5G
77159-850EA 42 USD
77159-850 77159-852 77159-854
(4-Bromobenzyl)hydrazine hydrochloride 95%
(4-Bromobenzyl)hydrazine hydrochloride

Precaución: for Research Use Only

Formula: C₇H₁₀BrClN₂
MDL Number: MFCD12031977
CAS Number: 1727-08-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR